Merck
Characterization of a liposome-based artificial skin membrane for in vitro permeation studies using Franz diffusion cell device.
Journal of liposome research (2016-09-02)
Hui Zhang, Xuemin Zhu, Jingjing Shen, Haiheng Xu, Min Ma, Wei Gu, Qiudong Jiang, Jun Chen, Jinao Duan
RÉSUMÉ
A prerequisite for successful transdermal or dermal drug therapy is the drug ability to penetration through the skin, especially stratum corneum (SC). The most acceptable technique for measuring skin permeation in vitro is the application of both the Franz diffusion cell device and the skin model. In the skin model, a liposome-based artificial skin membrane (LASM) consisting of tight layers of liposomes immobilized on a filter was prepared and characterized. Using porcine ear skin, rat skin and Strat-M™ artificial membrane as control, the LASM was then evaluated in permeation studies with five active compounds: ferulic acid, paeoniflorin, albiflorin, tetrahydrocolumbamine, and tetrahydropalmatine. The scanning electron microscope images demonstrated complete filling of the membrane pores with lipids and the formation of a continuous liposomal coating. The contents of egg phosphatidylcholine (EPC) and cholesterol in LASM were measured to be 12.08 ± 0.18 and 4.41 ± 0.04 mg/cm
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Albiflorin, ≥95% (LC/MS-ELSD)