Merck
  • Squamous cell carcinoma of the extrahepatic bile duct with metachronous para-aortic lymph node metastasis successfully treated with S-1 plus cisplatin.
Squamous cell carcinoma of the extrahepatic bile duct with metachronous para-aortic lymph node metastasis successfully treated with S-1 plus cisplatin.
BMJ case reports (2016-12-10)
Ryohei Nishiguchi, Dal Ho Kim, Masayuki Honda, Tsuguo Sakamoto
RÉSUMÉ
The most common histological classification of bile duct cancer is adenocarcinoma and squamous cell carcinoma (SCC) is relatively rare. We report a case of a 78-year-old man with SCC of the extrahepatic bile duct associated with metachronous para-aortic lymph node metastasis. He had undergone subtotal stomach-preserving pancreatoduodenectomy. The pathological findings demonstrated moderately differentiated SCC of the distal extrahepatic bile duct (T1N1M0, stage IIB). 6 months after surgery, recurrence of the para-aortic lymph node was shown in abdominal CT. 5 courses of tegafur/gimeracil/oteracil (S-1) plus cisplatin therapy was performed and the para-aortic lymph node disappeared, confirmed as complete response by imaging findings. The patient is alive without recurrence, 10 months after recurrence and chemotherapy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Gimeracil, ≥98% (HPLC)