Merck
Photoelectrochemical oxidation of organic substrates in organic media.
Nature communications (2017-09-01)
Tengfei Li, Takahito Kasahara, Jingfu He, Kevan E Dettelbach, Glenn M Sammis, Curtis P Berlinguette
RÉSUMÉ
There is a global effort to convert sunlight into fuels by photoelectrochemically splitting water to form hydrogen fuels, but the dioxygen byproduct bears little economic value. This raises the important question of whether higher value commodities can be produced instead of dioxygen. We report here photoelectrochemistry at a BiVO
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Vanadyl acetylacetonate, 98%