Merck
  • Kinetic and high-throughput profiling of epigenetic interactions by 3D-carbene chip-based surface plasmon resonance imaging technology.
Kinetic and high-throughput profiling of epigenetic interactions by 3D-carbene chip-based surface plasmon resonance imaging technology.
Proceedings of the National Academy of Sciences of the United States of America (2017-08-16)
Shuai Zhao, Mo Yang, Wenfei Zhou, Baichao Zhang, Zhiqiang Cheng, Jiaxin Huang, Min Zhang, Zhiyou Wang, Rui Wang, Zhonglei Chen, Jinsong Zhu, Haitao Li
RÉSUMÉ
Chemical modifications on histones and DNA/RNA constitute a fundamental mechanism for epigenetic regulation. These modifications often function as docking marks to recruit or stabilize cognate "reader" proteins. So far, a platform for quantitative and high-throughput profiling of the epigenetic interactome is urgently needed but still lacking. Here, we report a 3D-carbene chip-based surface plasmon resonance imaging (SPRi) technology for this purpose. The 3D-carbene chip is suitable for immobilizing versatile biomolecules (e.g., peptides, antibody, DNA/RNA) and features low nonspecific binding, random yet function-retaining immobilization, and robustness for reuses. We systematically profiled binding kinetics of 1,000 histone "reader-mark" pairs on a single 3D-carbene chip and validated two recognition events by calorimetric and structural studies. Notably, a discovery on H3K4me3 recognition by the DNA mismatch repair protein MSH6 in
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Bis[2-(2-bromoisobutyryloxy)undecyl] disulfide, 97%