Merck
IL-5 blocks apoptosis and tau hyperphosphorylation induced by Aβ
Journal of physiology and biochemistry (2017-01-31)
Yuanyuan Zhou, Chaoyan Li, Deheng Li, Yaping Zheng, Jin Wang
RÉSUMÉ
The primary features of Alzheimer's disease (AD) are extracellular amyloid plaques consisting mainly of deposits of amyloid β (Aβ) peptides and intracellular neurofibrillary tangles (NFTs). Sets of evidence suggest that interleukin-5 (IL-5) is involved in the pathogenesis of AD. Herein, we investigated the protective role of IL-5 in PC12 cells, to provide new insights into understanding this disease. Western blot was employed to assess the protein levels of Bax and phospho-tau as well as phospho-JAK2; MTT assay was performed to decipher cell viability. Treatment of IL-5 decreased Aβ
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human JAK2