Merck
T
Nature (2017-07-13)
Ilenia Papa, David Saliba, Maurilio Ponzoni, Sonia Bustamante, Pablo F Canete, Paula Gonzalez-Figueroa, Hayley A McNamara, Salvatore Valvo, Michele Grimbaldeston, Rebecca A Sweet, Harpreet Vohra, Ian A Cockburn, Michael Meyer-Hermann, Michael L Dustin, Claudio Doglioni, Carola G Vinuesa
RÉSUMÉ
Protective high-affinity antibody responses depend on competitive selection of B cells carrying somatically mutated B-cell receptors by follicular helper T (T
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, crystalline