- Gold nanorod embedded large-pore mesoporous organosilica nanospheres for gene and photothermal cooperative therapy of triple negative breast cancer.
Gold nanorod embedded large-pore mesoporous organosilica nanospheres for gene and photothermal cooperative therapy of triple negative breast cancer.
Nanoscale (2017-01-10)
Qianqian Ni, Zhaogang Teng, Meng Dang, Ying Tian, Yunlei Zhang, Peng Huang, Xiaodan Su, Nan Lu, Zhenlu Yang, Wei Tian, Shouju Wang, Wenfei Liu, Yuxia Tang, Guangming Lu, Longjiang Zhang
PMID28066849
RÉSUMÉ
To date, clinicians still lack an effective strategy to treat triple negative breast cancer (TNBC). In this work, we design for the first time a gold nanorod embedded large-pore mesoporous organosilica (GNR@LPMO) nanoplatform for gene and photothermal cooperative therapy of TNBC. The synthesized GNR@LPMOs possess a uniform size (175 nm), high surface area (631 m