Merck
Preserving π-conjugation in covalently functionalized carbon nanotubes for optoelectronic applications.
Nature communications (2017-01-31)
Antonio Setaro, Mohsen Adeli, Mareen Glaeske, Daniel Przyrembel, Timo Bisswanger, Georgy Gordeev, Federica Maschietto, Abbas Faghani, Beate Paulus, Martin Weinelt, Raul Arenal, Rainer Haag, Stephanie Reich
RÉSUMÉ
Covalent functionalization tailors carbon nanotubes for a wide range of applications in varying environments. Its strength and stability of attachment come at the price of degrading the carbon nanotubes sp
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2,3,3-Trimethylindolenine, 98%
Sigma-Aldrich
2-Hydroxy-5-nitrobenzaldehyde, 98%