Merck
Chronic exposure to endosulfan induces inflammation in murine colon via β-catenin expression and IL-6 production.
Journal of immunotoxicology (2016-08-06)
Martha Cecilia Téllez-Bañuelos, Jesse Haramati, Karina Franco-Topete, Jorge Peregrina-Sandoval, Ramon Franco-Topete, Galina P Zaitseva
RÉSUMÉ
Endosulfan (ENDO) is a widely used organochlorine (OC) pesticide and persistent organo-pollutant. Epidemiological studies have shown that high levels of OC exposure were related to colorectal cancer (CRC) incidence. The objectives of the present study were to evaluate histological changes in the colon, as well as in in situ expression of β-catenin and P-selectin, and serum levels of select pro-inflammatory cytokines in mice administered ENDO; there is a relationship between increased serum IL-6 and P-selectin levels in CRC patients and aberrant β-catenin signaling is important in initiation/maintenance of most CRCs. Mice were exposed to ENDO (at dose < LD50) orally once a week for up to 24 weeks, and monitored (inclusive) for a total of 42 weeks. The experiment was comprised of three groups, one that did not receive ENDO (olive oil vehicle), one administered 2 mg ENDO/kg/week and a positive control (for induction of CRC) given a weekly 20 mg 1,2-dimethylhydrazine (DMH)/kg injection. The results indicated that oral administration of ENDO provoked moderate inflammation starting at six weeks, and severe colonic inflammation with an appearance of dysplastic formations (aberrant crypts) in mice treated with ENDO (or DMH) for 12 weeks or longer. Serum IL-6 levels significantly increased starting at six weeks and rose to a peak of 15-fold higher than in controls at 42 weeks; TNFα levels likewise significantly increased, with a later peak (≈four-fold higher than controls) at 30-42 weeks. Immunohistochemical analysis of the colon also showed that expression of β-catenin and P-selectin increased with length of exposure to ENDO. Taken together, the results indicate that continued repeated oral exposure to ENDO induces increased expression of β-catenin and P-selectin, inflammation in the colon, and, ultimately, local tissue dysplasia.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
5,5-Dimethylhydantoin, 97%