Merck
  • A specific epitope of protein inhibitor of activated STAT3 is responsible for the induction of apoptosis in rat transformed mast cells.
A specific epitope of protein inhibitor of activated STAT3 is responsible for the induction of apoptosis in rat transformed mast cells.
Journal of immunology (Baltimore, Md. : 1950) (2009-02-10)
Zohar Yagil, Gillian Kay, Hovav Nechushtan, Ehud Razin
RÉSUMÉ
Protein inhibitor of activated STAT3 (PIAS3) functions in vivo as a key molecule in suppressing the transcriptional activity of both microphthalmia transcription factor (MITF) and STAT3, two transcription factors that play a major role in the development, phenotypic expression, and survival of mast cells and melanocytes. In the present study we have investigated the role played by PIAS3 in the regulation of cell cycle in mast cells and melanocytes. We have characterized the biological role of a 23-aa domain derived from PIAS3 that induces apoptosis in these cells by inhibiting the transcriptional activity of both MITF and STAT3. This PIAS3 inhibitor peptide could serve as the beginning of an in depth study for the development of peptide inhibitors for MITF and STAT3.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps monoclonal anti-PIAS3 antibody produced in mouse, ~2 mg/mL, clone PIA3, purified immunoglobulin, buffered aqueous solution