Merck
Expression of IRAK1 in lung cancer tissues and its clinicopathological significance: a microarray study.
International journal of clinical and experimental pathology (2015-01-01)
Xiuling Zhang, Yiwu Dang, Ping Li, Minhua Rong, Gang Chen
RÉSUMÉ
The interleukin-1 receptor associated kinases 1 (IRAK1) is a down stream effector molecule of the toll like receptor (TLR) signaling pathway, which is involved in inflammation, autoimmunity and cancer. However, the role of IRAK1 in lung cancer remains unclarified. Herein, we investigated the protein expression and the clinicopathological significance of IRAK1 in 3 formalin-fixed paraffin-embedded lung cancer tissue microarrays by using immunohistochemistry, which included 365 tumor and 30 normal lung tissues. We found that the expression of IRAK1 in lung cancer was significantly higher compared with that in normal lung tissues (P=0.002). Receiver operating characteristic (ROC) curves were generated to evaluate the power of IRAK1 to distinguish lung cancer from non-cancerous lung tissue. The area under curve (AUC) of ROC of IRAK1 was 0.643 (95% CI 0.550~0.735, P=0.009). Additionally, IRAK1 expression was related to clinical TNM stage (r=0.241, P < 0.001), lymph node metastasis (r=0.279, P < 0.001) and tumor size (r=0.299, P < 0.001) in lung cancer. In the subgroup of non-small cell lung cancer (NSCLC) and small cell lung cancer (SCLC), the positive rates of IRAK1 were both higher than that in the normal lung tissues (P=0.003, P=0.002, respectively). Further spearman analysis showed that IRAK1 protein in NSCLC was positive correlated with clinical TNM stage (r=0.222, P < 0.001), lymph node metastasis (r=0.277, P < 0.001), tumor size (r=0.292, P < 0.001) and distal metastasis (r=0.110, P=0.043). In conclusion, the expression of IRAK1 protein might be valuable in identifying patients with increased risks of lung cancer and might act as a target for diagnosis and gene therapy for lung cancer.