Merck
Multidisciplinary Comprehensive Care for Early Recommencement of Oral Intake in Older Adults With Severe Pneumonia.
Journal of gerontological nursing (2016-09-27)
Tamami Koyama, Hiroshi Shamoto, Hideaki Anzai, Yutaka Koganei, Keisuke Maeda, Hidetaka Wakabayashi
RÉSUMÉ
HOW TO OBTAIN CONTACT HOURS BY READING THIS ARTICLE INSTRUCTIONS 1.3 contact hours will be awarded by Villanova University College of Nursing upon successful completion of this activity. A contact hour is a unit of measurement that denotes 60 minutes of an organized learning activity. This is a learner-based activity. Villanova University College of Nursing does not require submission of your answers to the quiz. A contact hour certificate will be awarded once you register, pay the registration fee, and complete the evaluation form online at http://goo.gl/gMfXaf. To obtain contact hours you must: 1. Read the article, "Multidisciplinary Comprehensive Care for Early Recommencement of Oral Intake in Older Adults With Severe Pneumonia" found on pages 21-29, carefully noting any tables and other illustrative materials that are included to enhance your knowledge and understanding of the content. Be sure to keep track of the amount of time (number of minutes) you spend reading the article and completing the quiz. 2. Read and answer each question on the quiz. After completing all of the questions, compare your answers to those provided within this issue. If you have incorrect answers, return to the article for further study. 3. Go to the Villanova website listed above to register for contact hour credit. You will be asked to provide your name; contact information; and a VISA, MasterCard, or Discover card number for payment of the $20.00 fee. Once you complete the online evaluation, a certificate will be automatically generated. This activity is valid for continuing education credit until September 30, 2019. CONTACT HOURS This activity is co-provided by Villanova University College of Nursing and SLACK Incorporated. Villanova University College of Nursing is accredited as a provider of continuing nursing education by the American Nurses Credentialing Center's Commission on Accreditation. ACTIVITY OBJECTIVES 1. Describe the effect of multidisciplinary comprehensive care (MDCC) on oral intake in older adults with severe pneumonia at the time of discharge. 2. Explore the impact of MDCC on the length of stay for hospitalized older adults with severe pneumonia. DISCLOSURE STATEMENT Neither the planners nor the authors have any conflicts of interest to disclose. The current study was designed to assess the effect of multidisciplinary comprehensive care (MDCC) on (a) oral intake at discharge and (b) hospital stay duration in older adult patients with severe pneumonia. Participants were divided into two groups: receiving and not receiving MDCC. MDCC comprises regular assessment of swallowing ability, aspiration risk management, improvement of oral hygiene, serving of nutritious texture-modified foods, and encouragement of early mobilization. The MDCC group (164 women, 206 men; mean age = 82.7, SD = 8.4 years) had severe pneumonia as well as high proportions of poor premorbid physical function and consciousness disturbance compared to the non-MDCC group (45 women, 56 men; mean age = 81.1, SD = 8.6 years). Nevertheless, MDCC was an independent determinant of hospital stay duration and oral intake (Functional Oral Intake Scale score ≥4) at discharge with Cox regression analysis (hazard ratio = 1.42, 95% confidence interval [1.09, 1.85]). MDCC may promote early oral intake and hospital discharge in older adults with severe pneumonia. [Journal of Gerontological Nursing, 42(10), 21-29.].
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Guanosine 5′-monophosphomorpholidate 4-morpholine-N,N′-dicyclohexylcarboxamidine salt, ≥90%