Merck
Comparative Effects of Metal-Catalyzed Oxidizing Systems on Carbonylation and Integrity of Therapeutic Proteins.
Pharmaceutical research (2015-10-27)
Dmitry Kryndushkin, V Ashutosh Rao
RÉSUMÉ
Ascorbic acid has been considered as a potential radical scavenging excipient for pharmaceutical formulations. However, under certain circumstances, ascorbic acid can generate reactive oxygen species via redox cycling. The objective of this study was to investigate ascorbic acid-induced oxidative carbonylation of therapeutic proteins and correlate the increase in carbonylation with protein aggregation. An optimized ELISA for quantifying carbonyl levels was used to compare the oxidizing potentials of ascorbic acid and hydrogen peroxide by testing four pharmaceutically-relevant proteins (human serum albumin, immunoglobulin G, granulocyte-colony stimulating factor and calcitonin). Several transition metals at micromolar concentrations were evaluated for their ability to enhance ascorbic acid-induced protein carbonylation. Protein aggregation under oxidative conditions, with or without free radical scavengers, was measured by aggregate binding fluorescent dye and confirmed by microfluidic imaging. Addition of ascorbic acid alone resulted in higher increases in carbonylation than addition of hydrogen peroxide. The presence of trace amounts (>75 ppb) of copper enhanced oxidative effects of ascorbic acid, whereas other tested metals did not comparably promote oxidation. During oxidation, protein destabilization indicated by loss of the full-length protein, positively correlated with the increase in protein aggregation. However, levels of aggregation did not always correlate with the levels of protein carbonylation. At comparable carbonylation levels, addition of copper produced greater protein destabilization and aggregation than addition of iron. The results strongly suggest that ascorbic acid with traces of metals, especially copper, can promote therapeutic protein carbonylation and potentially aggregation. At similar carbonylation levels, some oxidative conditions may lead to greater protein destabilization than others.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Albumine human, recombinant, expressed in Saccharomyces cerevisiae, aqueous solution, 10% in aqueous buffer, ≥99% (agarose gel electrophoresis)