- Total synthesis of (S)-equol.
Total synthesis of (S)-equol.
Organic letters (2006-11-17)
Jennifer M Heemstra, Sean A Kerrigan, Daniel R Doerge, William G Helferich, William A Boulanger
PMID17107042
RÉSUMÉ
The first enantioselective total synthesis of (S)-equol is reported. The described route relies on an Evans alkylation to form the stereocenter and an intramolecular Buchwald etherification to generate the chroman ring. Key features of this method include its brevity, its scalability, and the low cost of starting materials. [reaction: see text].
MATÉRIAUX