Merck
Timosaponin-BII inhibits the up-regulation of BACE1 induced by ferric chloride in rat retina.
BMC complementary and alternative medicine (2012-10-23)
Ju-Fang Huang, Lei Shang, Pei Liu, Meng-Qi Zhang, Shuang Chen, Dan Chen, Chun-Ling Fan, Hui Wang, Kun Xiong
RÉSUMÉ
Our previous studies indicated that oxidative stress up-regulated the expression of β-amyloid precursor protein cleavage enzyme-1 (BACE1) in rat retina. Pharmacological reports have shown Timosaponin-BII, a purified extract originating from Chinese medical herb Rhizoma Anemarrhenae, is characterized as an antioxidant. Our present study aimed to determine whether Timosaponin-BII affected the expression of BACE1, β-amyloid precursor protein cleavage production of Aβ1-40 and β-C-terminal fragment (β-CTF) in rat retina, which were pre-treated with the oxidizing agent (solution of FeCl₃). Few distinctions of BACE1 distribution were observed among all groups (normal control group, model group, Timosaponin-BII treated and vehicle control groups). Rat retinas in model group and vehicle control group manifested an apparent up-regulation of BACE1 expression. Meanwhile, the level of malonaldehyde (MDA), Aβ1-40 and β-CTF were increased. However, when comparing with the vehicle control group, the retinas in Timosaponin-BII treated group showed significantly less BACE1 (p<0.05) and accumulated less Aβ1-40 or β-CTF (p<0.05). It also showed significantly decreased level of MDA (p<0.05) and prolonged partial thromboplastin time (p<0.05). Our data suggested that Timosaponin-BII remarkably inhibited the up-regulation of BACE1 and reduced the over-production of β-CTF and Aβ in rat retina, which was induced by FeCl₃. The mechanism of Timosaponin-BII on BACE1 expression may be related to its antioxidant property.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-BACE Antibody, CT, Chemicon®, from rabbit