Merck
Highly stereoselective hydrocarbation of terminal alkynes via Pt-catalyzed hydrosilylation/Pd-catalyzed cross-coupling reactions.
Organic letters (2001-03-30)
S E Denmark, Z Wang
RÉSUMÉ
[reaction: see text]. The formal addition of an aryl-H or alkenyl-H bond across a terminal alkyne has been accomplished by the combination of platinum-catalyzed hydrosilylation followed by palladium-catalyzed cross-coupling. The use of the t-Bu3P-Pt(DVDS) catalyst in combination with tetramethyldisiloxane gave excellent regio- and stereoselectivity with a number of alkyne substrates. Subsequent, fluoride-promoted cross-coupling proceeded in high yield and stereospecificity for a variety of aryl halides.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Complexe platine(0)-1,3-divinyl-1,1,3,3-tétraméthyldisiloxane solution, in xylene, Pt ~2 %