Merck
  • The effect of diabetes-associated autoantigens on cell processes in human PBMCs and their relevance to autoimmune diabetes development.
The effect of diabetes-associated autoantigens on cell processes in human PBMCs and their relevance to autoimmune diabetes development.
Journal of diabetes research (2013-07-11)
Jana Vcelakova, Radek Blatny, Zbynek Halbhuber, Michal Kolar, Ales Neuwirth, Lenka Petruzelkova, Tereza Ulmannova, Stanislava Kolouskova, Zdenek Sumnik, Pavlina Pithova, Maria Krivjanska, Dominik Filipp, Katerina Stechova
RÉSUMÉ
Type 1 Diabetes (T1D) is considered to be a T-helper- (Th-) 1 autoimmune disease; however, T1D pathogenesis likely involves many factors, and sufficient tools for autoreactive T cell detection for the study of this disease are currently lacking. In this study, using gene expression microarrays, we analysed the effect of diabetes-associated autoantigens on peripheral blood mononuclear cells (PBMCs) with the purpose of identifying (pre)diabetes-associated cell processes. Twelve patients with recent onset T1D, 18 first-degree relatives of the TD1 patients (DRL; 9/18 autoantibody positive), and 13 healthy controls (DV) were tested. PBMCs from these individuals were stimulated with a cocktail of diabetes-associated autoantigens (proinsulin, IA-2, and GAD65-derived peptides). After 72 hours, gene expression was evaluated by high-density gene microarray. The greatest number of functional differences was observed between relatives and controls (69 pathways), from which 15% of the pathways belonged to "immune response-related" processes. In the T1D versus controls comparison, more pathways (24%) were classified as "immune response-related." Important pathways that were identified using data from the T1D versus controls comparison were pathways involving antigen presentation by MHCII, the activation of Th17 and Th22 responses, and cytoskeleton rearrangement-related processes. Genes involved in Th17 and TGF-beta cascades may represent novel, promising (pre)diabetes biomarkers.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
L-Glutamine–Penicillin–Streptomycin solution, with 200 mM L-glutamine, 10,000 U penicillin and 10 mg steptomycin/mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture