Merck
Nurr1 regulates Top IIβ and functions in axon genesis of mesencephalic dopaminergic neurons.
Molecular neurodegeneration (2012-02-03)
Xin Heng, Gang Jin, Xin Zhang, Dehuang Yang, Mingzhe Zhu, Shijun Fu, Xuping Li, Weidong Le
RÉSUMÉ
NURR1 (also named as NR4A2) is a member of the steroid/thyroid hormone receptor family, which can bind to DNA and modulate expression of target genes. Previous studies have shown that NURR1 is essential for the nigral dopaminergic neuron phenotype and function maintenance, and the defects of the gene are possibly associated with Parkinson's disease (PD). In this study, we used new born Nurr1 knock-out mice combined with Affymetrix genechip technology and real time polymerase chain reaction (PCR) to identify Nurr1 regulated genes, which led to the discovery of several transcripts differentially expressed in the nigro-striatal pathway of Nurr1 knock-out mice. We found that an axon genesis gene called Topoisomerase IIβ (Top IIβ) was down-regulated in Nurr1 knock-out mice and we identified two functional NURR1 binding sites in the proximal Top IIβ promoter. While in Top IIβ null mice, we saw a significant loss of dopaminergic neurons in the substantial nigra and lack of neurites along the nigro-striatal pathway. Using specific TOP II antagonist ICRF-193 or Top IIβ siRNA in the primary cultures of ventral mesencephalic (VM) neurons, we documented that suppression of TOP IIβ expression resulted in VM neurites shortening and growth cones collapsing. Furthermore, microinjection of ICRF-193 into the mouse medial forebrain bundle (MFB) led to the loss of nigro-striatal projection. Taken together, our findings suggest that Top IIβ might be a down-stream target of Nurr1, which might influence the processes of axon genesis in dopaminergic neurons via the regulation of TOP IIβ expression. The Nurr1-Top IIβ interaction may shed light on the pathologic role of Nurr1 defect in the nigro-striatal pathway deficiency associated with PD.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human NR4A2