Merck
  • Surfactant protein-D attenuates the lipopolysaccharide-induced inflammation in human intestinal cells overexpressing toll-like receptor 4.
Surfactant protein-D attenuates the lipopolysaccharide-induced inflammation in human intestinal cells overexpressing toll-like receptor 4.
Pediatric surgery international (2015-10-30)
Ryuta Saka, Tetsu Wakimoto, Fumiko Nishiumi, Takashi Sasaki, Satoko Nose, Masahiro Fukuzawa, Takaharu Oue, Itaru Yanagihara, Hiroomi Okuyama
RÉSUMÉ
Necrotizing enterocolitis (NEC) is a devastating inflammatory disease of preterm infants that may depend on overexpression of toll-like receptor-4 (TLR4) in the immature intestine. Surfactant protein (SP)-D is a member of the collectin family and plays an important role in innate immunity, particularly in the airways. Although SP-D also exists in the intestines, little is known about its function. This study investigated whether SP-D can attenuate the inflammatory response of TLR4-overexpressing embryonal intestinal cells. All experimental procedures were performed using the human intestinal cell line INT407 originally derived from human embryonal intestines. Platelet-activating factor (PAF), reported to be elevated in NEC patients, was used to induce TLR4 overexpression in the human embryonal intestinal cell line INT407. TLR4 expression was measured using quantitative real-time PCR. Inflammatory responses to PAF (5 µM), the TLR4 agonist lipopolysaccharide (LPS, 100 ng/ml), PAF + LPS, and PAF + LPS following SP-D pretreatment (20 µg/ml) were assessed by enzyme-linked immunosorbent assay (ELISA) of interleukin-8 (IL-8) release (in pg/ml). Expression of TLR4 mRNA (mean ± SD) was upregulated by PAF (369 % ± 28 %, p < 0.001). Stimulation with PAF + LPS resulted in higher IL-8 release (1959.3 ± 52.3) than control (141.2 ± 12.4), LPS (167.3 ± 65.8), or PAF (1527.2 ± 129.4) treatment (p < 0.05). Release in response to PAF + LPS (1590.1 ± 319.3) was attenuated by SP-D pretreatment (1161.6 ± 131.6; p < 0.05). SP-D attenuates LPS-induced IL-8 production in TLR4-overexpressing intestinal cells, suggesting that SP-D may have a protective effect in the development of NEC in preterm infants.