Merck
Activation of retinal rod cGMP-gated channels: what makes for an effective 8-substituted derivative of cGMP?
Biochemistry (1993-09-28)
R L Brown, R J Bert, F E Evans, J W Karpen
RÉSUMÉ
Analogs of cGMP bearing diverse substituents at the C8 position of the guanine ring system have been shown to activate the cGMP-activated channel of retinal rods at concentrations lower than cGMP itself. In an effort to understand this behavior, we synthesized eight novel C8-substituted derivatives and tested their ability to activate channels in excised patches from salamander rod outer segments. We express the effectiveness of each analog as a ratio (in brackets) of the concentration required to open half of the channels in a patch to that required of 8-Br-cGMP, previously shown to be about 10 times more effective than cGMP. Five of the derivatives contained a thio substitution at C8: n-propylthio-cGMP [0.61], sulfoethylthio-cGMP [0.90], carboxyethylthio-cGMP [0.97], aminoethylthio-cGMP [2.8], and (trimethylamino)ethylthio-cGMP [8.5]. Three of the derivatives contained an amino substitution at C8: carboxyethylamino-cGMP [22], n-propylamino-cGMP [25], and aminoethylamino-cGMP [230]. The results indicate that thio-substitution at C8 produces more effective analogs than does amino-substitution, regardless of the chemical nature of the terminal functional group. Derivatives containing neutral and apolar tails opened channels at much lower concentrations than their positively-charged counterparts with the same C8 substituent. Analogs having negatively-charged tails were also more effective than those with positive charge but not quite as effective as those with neutral tails.(ABSTRACT TRUNCATED AT 250 WORDS)
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-Bromopropionic acid, 97%