Merck
CFTR regulates outwardly rectifying chloride channels through an autocrine mechanism involving ATP.
Cell (1995-06-30)
E M Schwiebert, M E Egan, T H Hwang, S B Fulmer, S S Allen, G R Cutting, W B Guggino
RÉSUMÉ
The cystic fibrosis transmembrane conductance regulator (CFTR) functions to regulate both Cl- and Na+ conductive pathways; however, the cellular mechanisms whereby CFTR acts as a conductance regulator are unknown. CFTR and outwardly rectifying Cl- channels (ORCCs) are distinct channels but are linked functionally via an unknown regulatory mechanism. We present results from whole-cell and single-channel patch-clamp recordings, short-circuit current recordings, and [gamma-32P]ATP release assays of normal, CF, and wild-type or mutant CFTR-transfected CF airway cultured epithelial cells wherein CFTR regulates ORCCs by triggering the transport of the potent agonist, ATP, out of the cell. Once released, ATP stimulates ORCCs through a P2U purinergic receptor-dependent signaling mechanism. Our results suggest that CFTR functions to regulate other Cl- secretory pathways in addition to itself conducting Cl-.