Merck
  • Inhibition of reverse-mode sodium-calcium exchanger activity and apoptosis by levosimendan in human cardiomyocyte progenitor cell-derived cardiomyocytes after anoxia and reoxygenation.
Inhibition of reverse-mode sodium-calcium exchanger activity and apoptosis by levosimendan in human cardiomyocyte progenitor cell-derived cardiomyocytes after anoxia and reoxygenation.
PloS one (2014-02-06)
Ping-Chun Li, Ya-Chi Yang, Guang-Yuh Hwang, Lung-Sen Kao, Ching-Yuang Lin
RÉSUMÉ
Levosimendan, a known calcium sensitizer with positive inotropic and vasodilating properties, might also be cardioprotective during ischemia-reperfusion (I/R) insult. Its effects on calcium homeostasis and apoptosis in I/R injury remain unclear. Na(+)/Ca(2+) exchanger (NCX) is a critical mediator of calcium homeostasis in cardiomyocytes, with reverse-mode NCX activity responsible for intracellular calcium overload and apoptosis of cardiomyocytes during I/R. We probed effects and underlying mechanisms of levosimendan on apoptosis and NCX activity in cultured human cardiomyocyte progenitor cells (CPC)-derived cardiomyocytes undergoing anoxia-reoxygenation (A/R), simulating I/R in vivo. Administration of levosimendan decreased apoptosis of CPC-derived cardiomyocytes induced by A/R. The increase in reverse-mode NCX activity after A/R was curtailed by levosimendan, and NCX1 was translocated away from the cell membrane. Concomitantly, endoplasmic reticulum (ER) stress response induced by A/R was attenuated in CPC-derived cardiomycytes treated with NCX-targeted siRNA or levosimendan, with no synergistic effect between treatments. Results indicated levosimendan inhibited reverse-mode NCX activity to protect CPC-derived cardiomyocytes from A/R-induced ER stress and cell death.