Merck
Recent advances in the Willgerodt-Kindler reaction.
Chemical Society reviews (2013-06-25)
Daniel L Priebbenow, Carsten Bolm
RÉSUMÉ
The Willgerodt-Kindler reaction has, in recent years, received limited attention and application in synthetic organic chemistry. With the advent of new technology such as microwave-assisted heating, several new, high-yielding, practical, and more environmentally friendly reaction protocols have been developed. This review aims to once again draw attention to this relatively underutilised process by highlighting the recent developments in the Willgerodt-Kindler reaction in the synthesis of (thio)amides, carboxylic acids, and heterocycles.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Sodium sulfide nonahydrate, ACS reagent, ≥98.0%