Merck
Intravenous fenoldopam for early acute kidney injury after liver transplantation.
Journal of anesthesia (2014-12-01)
Gianni Biancofiore, Maria L Bindi, Mario Miccoli, Elisabetta Cerutti, Bruna Lavezzo, Laura Pucci, Massimo Bisà, Massimo Esposito, Luca Meacci, Roberto Mozzo, Chiara Stratta, Giuseppe Penno, Angelo Baggiani, Franco Filipponi
RÉSUMÉ
Acute kidney injury remains a serious complication after orthotopic liver transplantation. To date, several 'renal-protective' agents have been explored in this setting but with conflicting and disappointing results. Therefore, our aim is to evaluate the effects of fenoldopam in liver transplant patients with an established renal injury. In this prospective study, intravenous fenoldopam 0.1 µg/kg/min was administered to consecutive liver transplant patients with postoperative (within 7 days from surgery) stage 2 acute kidney injury (AKI) according to the Acute Kidney Injury Network classification. Actual glomerular filtration rate (GFR; calculated by the iohexol plasma clearance), serum creatinine (SCr) and cystatin C (SCyC) were used to assess the effect of the medication on the patients. During the study, 295 patients underwent liver transplant. Fifty-one patients (17.6%) met the inclusion criteria and the data from 48 patients were analysed. SCr and SCyC levels decreased (p < 0.001 after 48 h; p < 0.0001 after 72 h) and GFR increased (p < 0.001 after 24 h; p < 0.0001 after 72 h). When compared to a cohort of comparable patients with AKI from our historical series, the patients in the present study showed better SCr and SCyC levels. It was not necessary to discontinue the infusion of fenoldopam in any patient because of the occurrence of adverse events potentially attributable to it. We showed that fenoldopam was capable of improving some renal function parameters in postoperative liver transplantation patients with on-going AKI. This preliminary study now sets the stage for a multicenter, randomized, placebo-controlled trial in order to provide definite evidence.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme
Sigma-Aldrich
Iode, ≥99.99% trace metals basis
Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme
Sigma-Aldrich
Prednisolone, ≥99%
Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme
Sigma-Aldrich
Iode, 99.999% trace metals basis
Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme
Sigma-Aldrich
Iode, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, for molecular biology, ≥95%
Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme
Sigma-Aldrich
FK-506 monohydrate, ≥98% (HPLC)
Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme