Merck
Enhancement of solubility and dissolution of cilostazol by solid dispersion technique.
Archives of pharmacal research (2015-01-09)
Jun-Hyung Park, Hoo-Kyun Choi
RÉSUMÉ
Cilostazol is practically insoluble in water and thus results in poor bioavailability. Only a few approaches have been reported for improving the bioavailability of cilostazol. Solid dispersion technique via solvent evaporation method was applied to improve the solubility and dissolution of cilostazol. Various polymers, mixture of polymer and surfactant, and mixture of polymers were screened as a carrier for the solid dispersion. Solubility of cilostazol was improved significantly when Eudragit(®) L100 was used as a carrier. However, addition of surfactant to Eudragit(®) L100 decreased the solubility slightly. Whereas, the mixture of Eudragit(®) L100 and Eudragit(®) S100 as a carrier system further increased the solubility. Based on the highest solubility obtained among the carriers screened, 1:1 ratio of Eudragit(®) L100 and Eudragit(®) S100 was selected as a carrier, and drug to carrier ratio was optimized to 1:5. Differential scanning calorimetry and X-ray diffraction studies showed that the characteristic peak of cilostazol disappeared in the solid dispersion, indicating that cilostazol existed in amorphous form in this formulation. Spray drying method was superior to vacuum drying method in terms of dissolution rate. Meanwhile, it was observed that the disintegration rate and the concentration of polymer had some effect on the crystallization of cilostazol in dissolution medium. Tablet formulation containing spray dried solid dispersion showed significant improvement in dissolution as compared to the commercial tablet.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Vinyl acetate, contains 3-20 ppm hydroquinone as inhibitor, ≥99%
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Le tarif et la disponibilité ne sont pas disponibles actuellement.