Merck
A new irreversible enzyme-aided esterification method in organic solvents.
Biotechnology letters (2008-01-16)
Günter E Jeromin, Annegreth Zoor
RÉSUMÉ
A new irreversible esterification method for carboxylic acids catalyzed by a lipase from Candida antarctica (Novozyme 435) in organic solvents has been developed. The water produced during the process is chemically destroyed by a corresponding ester of acetoacetate, which acts as a sacrificial substrate in this reaction. The flavour esters isobutyl acetate, methyl butyrate, ethyl butyrate and benzyl butyrate were synthesized either in small scale (0.05 mol) or large scale (1 mol). The yields range from 82 to 92% within 24 h at 52 degrees C. Optimal molar ratios of reactants were 1:1:1 (carboxylic acid:alcohol:acetoacetate).
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Isobutyl acetate, 99%