Merck
Adiponectin-induced antitumor activity on prostatic cancers through inhibiting proliferation.
Cell biochemistry and biophysics (2014-05-06)
Qiruo Gao, Junhua Zheng
RÉSUMÉ
Adiponectin, an adipose tissue-derived hormone, has been studied intensively for the past decade because of its anti-inflammatory, anti-atherogenic, and anti-diabetic properties. Recent advances suggest that adiponectin also plays an important role in the development and progression of various cancers. Accumulating evidence suggests that adiponectin may have an important protective role in carcinogenesis. Adiponectin circulates at high concentrations in human plasma. Plasma levels of adiponectin are approximately 50 % lower in obese than in lean subjects. An association between low plasma levels of adiponectin and higher risk of developing prostate and other cancers was recently reported. Obesity and overweight have also been associated with increased mortality from cancer. To test the hypothesis that adiponectin exerts direct antiproliferative and/or pro-apoptotic effects on cancer cells, we used the PC-3 human prostate adenocarcinoma cell line. The proliferation rate of the PC-3 cells was measured using the MTT method, and apoptosis was examined by quantifying the DNA fragmentation using an ELISA assay. In addition, adiponectin receptor 1 (AdipoR1) and AdipoR2 mRNA expression was detected using RT-PCR. Adiponectin diminished the proliferation rate of PC-3 cells; this effect was significant after 48-96 h of treatment. The presence of receptor expression suggested that the effect of adiponectin on cell proliferation was most likely specific and adiponectin receptor-mediated. Adiponectin induced no apoptosis of PC-3 cells over 48 h. We conclude that adiponectin inhibits proliferation but causes no apoptosis of PC-3 prostate cancer cells.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Boc-Asp(OMe)-fluoromethyl ketone, ≥90% (TLC), solid