Merck
Sheltering effect and indirect pathogenesis of carbapenem-resistant Acinetobacter baumannii in polymicrobial infection.
Antimicrobial agents and chemotherapy (2014-05-07)
Yu-Ting Liao, Shu-Chen Kuo, Yi-Tzu Lee, Chien-Pei Chen, Shu-Wen Lin, Li-Jiuan Shen, Chang-Phone Fung, Wen-Long Cho, Te-Li Chen
RÉSUMÉ
The role of carbapenem-resistant Acinetobacter baumannii (CRAb) in polymicrobial infection remains elusive. Having observed the ability of CRAb to shelter other susceptible bacteria from carbapenem killing, we sought to determine the factors contributing to this sheltering effect by transforming different recombinant plasmids into recipient A. baumannii cells. The sheltering effects of CRAb were reproduced in recipient A. baumannii cells that highly expressed carbapenem-hydrolyzing class D β-lactamases (CHDLs) through their associated strong promoter. With the use of Western blot analysis and a bioassay, the highly expressed CHDLs were found to be extracellularly released and led to hydrolysis of carbapenem. The level of extracellular CHDLs increased after challenge with a higher concentration of CHDL substrates, such as carbapenem and ticarcillin. This increased CHDL may, in part, be attributed to cell lysis, as indicated by the presence of extracellular gyrase. In the planktonic condition, the sheltering effect for the cocultured susceptible bacteria might represent an indirect and passive effect of the CRAb self-defense mechanism, because coculture with the susceptible pathogen did not augment the amount of the extracellular CHDLs. Polymicrobial infection caused by CRAb and a susceptible counterpart exerted higher pathogenicity than monomicrobial infection caused by either pathogen alone in mice receiving carbapenem therapy. This study demonstrated that CHDL-producing CRAb appears to provide a sheltering effect for carbapenem-susceptible pathogens via the extracellular release of CHDLs and, by this mechanism, can enhance the pathogenesis of polymicrobial infection in the presence of carbapenem therapy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Millipore
Ticarcillin Supplement, suitable for microbiology