Merck
MSCs inhibit bone marrow-derived DC maturation and function through the release of TSG-6.
Biochemical and biophysical research communications (2014-07-12)
Yi Liu, Zhilin Yin, Run Zhang, Ke Yan, Lei Chen, Fanfan Chen, Weiyi Huang, Bingke Lv, Chengmei Sun, Xiaodan Jiang
RÉSUMÉ
Dendritic cells (DCs) are potent antigen-presenting cells (APCs) that are characterized by the ability to take up and process antigens and prime T cell responses. Mesenchymal stem cells (MSCs) are multipotent cells that have been shown to have immunomodulatory abilities, including inhibition of DC maturation and function in vivo and in vitro; however, the underlying mechanism is far from clear. In this study we found that MSCs can inhibit the maturation and function of bone marrow-derived DCs by releasing TSG-6. In the presence of MSCs, lower expression of mature DC surface phenotype (CD80, CD86, MHC-II, and CD11c) was observed. In addition, typical DC functions, such as the production of IL-12 and the ability to prime T cells, were decreased when co-cultured with MSCs. In contrast, knockdown of TSG-6 reduced the inhibitory effect of MSCs on DC. Moreover, we found that TSG-6 can suppress the activation of MAPKs, and NF-κB signaling pathways within DCs during Lipopolysaccharides (LPS) stimulation. In conclusion, we suggest that TSG-6 plays an important role in MSCs-mediated immunosuppressive effect on DC.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Mitomycine C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Mitomycine C from Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Mitomycine C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Mitomycine C from Streptomyces caespitosus, meets USP testing specifications
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tnfaip6
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
MISSION® esiRNA, targeting human TNFAIP6
Référence
Conditionnement
Disponibilité
Prix
Quantité