Merck
  • Association of variation in the LAMA3 gene, encoding the alpha-chain of laminin 5, with atopic dermatitis in a German case-control cohort.
Association of variation in the LAMA3 gene, encoding the alpha-chain of laminin 5, with atopic dermatitis in a German case-control cohort.
BMC dermatology (2014-11-05)
Susanne Stemmler, Qumar Parwez, Elisabeth Petrasch-Parwez, Joerg T Epplen, Sabine Hoffjan
RÉSUMÉ
Atopic dermatitis (AD) is a chronic inflammatory skin disorder caused by complex interaction of genetic and environmental factors. Besides mutations in the filaggrin gene, leading to impaired skin barrier function, variation in genes encoding additional skin proteins has been suggested to contribute to disease risk. Laminin 5, playing an important role in skin integrity, is composed of three subunits encoded by the LAMA3, LAMB3 and LAMC2 genes in which biallelic mutations cause epidermolysis bullosa junctionalis. We aimed at evaluating the role of variation in the LAMA3, LAMB3 and LAMC2 genes for AD pathogenesis. 29 single nucleotide polymorphisms (SNPs) were genotyped in the three genes in a German AD case-control cohort comprising 470 unrelated AD patients and 320 non-atopic controls by means of restriction enzyme digestion. Allele, genotype and haplotype frequencies were compared between cases and controls using chi-square testing and the Haploview software. Several SNPs in the LAMA3 gene showed significant association with AD in our cohort (p <0.01), while we did not detect association with variations in the LAMB3 and LAMC2 genes. Haplotype analysis additionally revealed several significantly associated haplotypes in the LAMA3 gene. Due to extensive linkage disequilibrium, though, we were not able to further differentiate the specific disease causing variation(s) in this region. We established the LAMA3 gene as novel potential susceptibility gene for AD. Additional studies in independent cohorts are needed to replicate these results.