Merck
Gene analysis in patients with premature ovarian failure or gonadal dysgenesis: a preliminary study.
Maturitas (2007-06-15)
Ana Maria Massad-Costa, Ismael Dale Cotrim Guerreiro da Silva, Regina Affonso, José Maria Soares, Márcia Gaspar Nunes, Geraldo Rodrigues de Lima, Edmund C Baracat
RÉSUMÉ
The aim of this study was to evaluate the presence of mutations in the coding region of the QM gene and fragile X in patients with premature ovarian failure and gonadal dysgenesis. After approval by the local Ethics Committee, blood samples, in EDTA, of 100 normally ovulating women, 23 with premature ovarian failure (POF) and 14 with gonadal dysgenesis 46XX, aged less than 40 years, were screened for mutation in the QM gene coding region. All patients with POF have 46, XX karyotype and serum levels of follicle-stimulating hormone (FSH) over 30 mIU/mL. In addition, all samples from patients with premature ovarian failure underwent analysis for fragile X. The QM gene located at a hotspot region (Xq28) showed five points of mutations in a patient with premature ovarian failure. Four of them were able to change the amino acid sequence of the protein. None of our patients were diagnosed as having pre or mutant X fragile syndrome. Our study suggests that Xq28 (QM gene) may be involved in ovary failure. However, further studies are needed to confirm this hypothesis.