Merck
  • The primary structure of human secretogranin II, a widespread tyrosine-sulfated secretory granule protein that exhibits low pH- and calcium-induced aggregation.
The primary structure of human secretogranin II, a widespread tyrosine-sulfated secretory granule protein that exhibits low pH- and calcium-induced aggregation.
The Journal of biological chemistry (1989-07-15)
H H Gerdes, P Rosa, E Phillips, P A Baeuerle, R Frank, P Argos, W B Huttner
RÉSUMÉ
Secretogranin II (previously also called chromogranin C) is a tyrosine-sulfated secretory protein found in secretory granules in a wide variety of endocrine cells and neurons. Here, we have determined the primary structure of human secretogranin II from a full length cDNA clone and have investigated its properties, predicted from the sequence, by studying the behavior of purified secretogranin II under conditions characteristic of the milieu of secretory granules. Analysis of a 2.35-kilobase cDNA clone isolated from a human pituitary library and identified as secretogranin II by various criteria showed that human presecretogranin II is a 617-residue polypeptide containing an NH2-terminal located signal peptide. Secretogranin II lacks the disulfide-bonded loop structure near the NH2 terminus which is conserved in chromogranin A and chromogranin B (secretogranin I), two other widespread constituents of neuroendocrine secretory granules, but like the latter two proteins contains (i) an -E-N/S-L-X-A/D-X-D/E-X-E-L- motif and (ii) multiple potential dibasic cleavage sites for the generation of smaller, perhaps biologically active peptides. Another structural feature that secretogranin II shares with chromogranin A and chromogranin B (secretogranin I) is the abundance of acidic residues all along the polypeptide chain whose negative charge must somehow be neutralized to allow condensation and packaging of the protein into secretory granules. Experiments with purified secretogranin II showed that in the presence of 10 mM calcium at pH 5.2, conditions characteristic of the milieu of neuroendocrine secretory granules, this protein formed aggregates. Immunoglobulin G, a secretory protein that in vivo is not packaged into secretory granules, did not form aggregates under these in vitro conditions and was excluded from the secretogranin II aggregates. Very little aggregation of secretogranin II was observed in the absence of calcium at pH 5.2 or in the presence of calcium at neutral pH. In vivo, ammonium chloride, which is known to neutralize the pH of acidic intracellular compartments, inhibited the packaging of newly synthesized secretogranin II into secretory granules. Our results suggest that the low pH- and calcium-induced aggregation of secretogranin II may be important for the organization of the secretory granule matrix and raise the possibility that aggregation of secretogranin II may be involved in its sorting to secretory granules.