Merck
Identification and in silico characterization of a novel gene: TPA induced trans-membrane protein.
Biochemical and biophysical research communications (2005-03-02)
Chung-Yip Chan, Mohammad R Salabat, Xian-Zhong Ding, David L Kelly, Mark S Talamonti, Richard H Bell, Thomas E Adrian
RÉSUMÉ
12-O-Tetradecanoylphorbol-13-acetate (TPA) is a potent tumor promoter with wide ranging, diverse, and sometimes opposite cellular effects. Using oligonucleotide microarray analysis, we have identified a novel gene that is upregulated following treatment with TPA in the pancreatic cancer cell line CD18. Real-time PCR validated the microarray results in CD18 and HeLa cells, and showed that upregulation of the gene is time- and concentration-dependent. In silico analysis showed the gene product to be a single-pass transmembrane protein of 217 residues that is localized to the endoplasmic reticulum, thus the name TPA induced trans-membrane protein (TTMP). A luciferase reporter assay demonstrated that upregulation of TTMP by TPA is triggered at the promoter level.