Merck
ZNF435, a novel human SCAN-containing zinc finger protein, inhibits AP-1-mediated transcriptional activation.
Molecules and cells (2007-07-25)
Xing Gu, Mei Zheng, Xiangwei Fei, Zhenxing Yang, Fan Li, Chaoneng Ji, Yi Xie, Yumin Mao
RÉSUMÉ
Zinc finger transcription factor genes are a significant fraction of the genes in the vertebrate genome. Here we report the isolation and characterization of a human zinc finger-containing gene, ZNF435, from a fetal brain cDNA library. ZNF435 cDNA is 1290 base pairs in length and contains an open reading frame encoding 349 amino acids with four C2H2-type zinc fingers at its carboxyl terminus and a SCAN motif at its amino terminus. RT-PCR results showed that ZNF435 was expressed in all tested tissues. A ZNF435-GFP fusion protein was located in the nucleus and the four zinc fingers acted as nuclear localization signals (NLSs). ZNF435 was found to be capable of homo-association, and this effect was independent of its zinc fingers. Furthermore, ZNF435 proved to be a transcription repressor as its overexpression in AD293 cells inhibited the transcriptional activities of AP-1.