Merck
Tungsten-catalyzed asymmetric epoxidation of allylic and homoallylic alcohols with hydrogen peroxide.
Journal of the American Chemical Society (2014-01-16)
Chuan Wang, Hisashi Yamamoto
RÉSUMÉ
A simple, efficient, and environmentally friendly asymmetric epoxidation of primary, secondary, tertiary allylic, and homoallylic alcohols has been accomplished. This process was promoted by a tungsten-bishydroxamic acid complex at room temperature with the use of aqueous 30% H2O2 as oxidant, yielding the products in 84-98% ee.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
α-Vinylbenzyl alcohol, 97%