Merck
Multifunctional receptor stabilin-1 in homeostasis and disease.
TheScientificWorldJournal (2010-10-19)
Julia Kzhyshkowska
RÉSUMÉ
The multifunctional scavenger receptor stabilin-1 (STAB1, FEEL-1, CLEVER-1, KIAA0246) is expressed on tissue macrophages and sinusoidal endothelial cells in healthy organisms, and its expression on both macrophages and different subtypes of endothelial cells is induced during chronic inflammation and tumor progression. Stabilin-1 is a type-1 transmembrane receptor that mediates endocytic and phagocytic clearance of "unwanted-self" components, intracellular sorting of the endogenously synthesized chitinase-like protein SI-CLP, and transcytosis of the growth hormone family member placental lactogen. The central sorting station for stabilin-1 trafficking seems to be the trans-Golgi network (TGN). Transport of stabilin-1 in the TGN requires interaction with GGA adaptors that bind to the classical DDSLL motif and a novel acidic cluster in its cytoplasmic tail. Degradation of stabilin-1 seems to depend on the interaction with sorting nexin 17. However, the mechanisms keeping stabilin-1 on the cell surface remain to be identified. This issue deserves specific attention due to the growing amount of data indicating that function of stabilin-1 in cell adhesion events is essential for inflammation and metastasis. Taking into consideration the complexity of stabilin-1-mediated processes, investigation of stabilin-1 functions in the animal models, as well as mathematic modeling of intracellular trafficking and extracellular contact, would enable prediction of stabilin-1 behavior in complex biological systems and would open perspectives for therapeutic targeting of stabilin-1 pathways in chronic inflammation and carcinogenesis.