Merck
Identification of urinary and intestinal bacterial metabolites of ellagitannin geraniin in rats.
Journal of agricultural and food chemistry (2008-01-01)
Hideyuki Ito, Ayumu Iguchi, Tsutomu Hatano
RÉSUMÉ
Hydrolyzable tannins, including ellagitannins, occur in foods such as berries and nuts. Various biological activities, including antioxidant, antiviral, and antitumor activities, have been noted and reported for ellagitannins, but the absorption and metabolism of purified ellagitannins are poorly understood. We describe herein the characterization of urinary and intestinal microbial metabolites in rats after the ingestion of ellagitannins. Urine samples were collected after oral administration of ellagitannins such as geraniin ( 1), corilagin ( 2), and their related polyphenols. The suspension of rat intestinal microflora was anaerobically incubated with ellagitannins. Each sample was separated by column chromatography and/or preparative HPLC to give seven metabolites, M1- M7. The structures of these metabolites were determined on the basis of spectroscopic data and chemical evidence. These compounds, except for M1, were characterized as ellagitannin metabolites for the first time. Furthermore, among four major metabolites ( M1- M4) in urine, M2 showed an antioxidant activity comparable to intact geraniin and related polyphenols.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
2-Bromo-4,5-dimethoxybenzoic acid, 98%