Merck
  • Macrophage migration inhibitory factor: association of -794 CATT5-8 and -173 G>C polymorphisms with TNF-α in systemic lupus erythematosus.
Macrophage migration inhibitory factor: association of -794 CATT5-8 and -173 G>C polymorphisms with TNF-α in systemic lupus erythematosus.
Human immunology (2014-02-18)
U De la Cruz-Mosso, R Bucala, C A Palafox-Sánchez, I Parra-Rojas, J R Padilla-Gutiérrez, A L Pereira-Suárez, H Rangel-Villalobos, M Vázquez-Villamar, L I Angel-Chávez, J F Muñoz-Valle
RÉSUMÉ
Macrophage migration inhibitory factor (MIF) is an upstream immunoregulatory cytokine associated with the pathogenesis of autoimmune inflammatory diseases. There is evidence that MIF functions in a positive feedback loop with TNF-α that could perpetuate the inflammatory process in systemic lupus erythematosus (SLE). In this case-control study we investigated whether commonly occurring functional MIF polymorphisms are associated with SLE as well as with MIF and TNF-α serum levels in a Mexican-Mestizo population. Genotyping of the -794 CATT5-8 (rs5844572) and -173 G>C (rs755622) MIF polymorphisms was performed by PCR and PCR-RFLP, respectively in 186 SLE patients and 200 healthy subjects. MIF and TNF-α serum levels were determined by ELISA. A significant increase of MIF and TNF-α levels was found in SLE patients. According to a genetic model, we found a significant association of genotypes carrying the -794 CATT7 and -173(∗)C risk alleles with susceptibility to SLE and with a significant increase of TNF-α. In conclusion, MIF gene polymorphisms are associated with SLE susceptibility and with an increase of TNF-α serum levels in a Mexican-Mestizo population.