Merck
Diagnostic utility of GLUT-1 expression in the cytologic evaluation of serous fluids.
Acta cytologica (2006-02-03)
Alaa Afify, Heidi Zhou, Lydia Howell, Augusto F G Paulino
RÉSUMÉ
To evaluate the extent to which adenocarcinomas in body cavity fluids express GLUT-1 in comparison to currently available markers for adenocarcinomas. Archival paraffin-embedded cell blocks of serous fluids from 25 cases of benign effusions containing reactive mesothelial cells and 39 cases of malignant effusions with metastatic adenocarcinoma (11 ovarian, 11 pulmonary, 9 gastrointestinal and 8 breast) were retrieved from the surgical pathology files. All cases were stained with antibodies for GLUT-1, Ber-Ep4, B72.3 and CEA. Positive staining was defined as distinct linear membrane staining for GLUT-1 and Ber-EP4, cytoplasmic staining for CEA, and cytoplasmic or membrane staining for B72.3. Strong staining in at least 10% of the tumor cells was required in order to consider the case positive for the particular marker. GLUT-1 was expressed in 72% (28 of 39) of cases of malignant effusions: 100% (11 of 11) from the ovary, 91% (10 of 11) from the lung, 67% (6 of 9) from the gastrointestinal tract and 12% (1 of 8) from the breast. None (0 of 25) of the benign effusions expressed GLUT-1. Malignant effusions expressed CEA in 74% (29 of 39), Ber-Ep4 in 85% (33 of 39), and B72.3 in 62% (24 of 39). Benign effusions expressed CEA in 3 cases and B72.3 in 2 cases. GLUT-1 is a useful marker that can be applied to cytologic specimens. It can be used as a reliable component of an antibody panel to distinguish reactive mesothelial cells from metastatic adenocarcinoma in particular adenocarcinomas of body cavity effusions, in particular adenocarcinomas of ovarian and pulmonary origin.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
GLUT1 Rabbit Polyclonal Antibody