Merck
Non-B, non-T neoplasms with lymphoblast morphology: further clarification and classification.
The American journal of surgical pathology (2003-09-26)
Kennosuke Karube, Koichi Ohshima, Takeshi Tsuchiya, Takahiro Yamaguchi, Hiroaki Suefuji, Junji Suzumiya, Mine Harada, Masahiro Kikuchi
RÉSUMÉ
We studied the morphologic, immunohistochemical, and clinical characteristics of 158 cases of lymphoblastic lymphoma. Based on immunophenotyping and cell lineage, cases were classified into B-cell type (CD20,CD19 or CD79a+, n = 53), T-cell type (surface CD3+, n = 84), and non-B, non-T type (B cell marker- and surface CD3-, n = 21). The latter group was further divided based on immunohistochemistry into: 1) CD7+ stem cell lymphoma (CD7+SCL) [CD4-, CD7+, CD33+/-, CD56-], 2) blastic natural killer cell lymphoma (B-NKL) [CD4-, CD7+/-, CD33-, CD56+, CD123-], 3) myeloid/NK precursor cell leukemia (M/NKL) [CD4-, CD7+, CD33+, CD56+], and 4) CD4+CD56+ hematodermic malignancy (CD4+CD56+) type [CD4+, CD7+/-, CD33-, CD56+, CD123+]. The CD7+SCL and M/NKL types frequently exhibited bone marrow invasion and mediastinal masses. All CD4+CD56+ types were associated with skin lesions. B-NKL type is included into Blastic NK lymphoma in new World Health Organization classification with CD4+CD56+ type. But the cases of B-NKL were more reminiscent of CD7+SCL or M/NKL type than the CD4+CD56+ type, both clinically and histologically. We propose that blastic NK lymphoma, a disease entity in the new WHO classification, should be divided into two types based on phenotypes and clinical features. The non-B, non-T lymphomas exhibited poorer prognoses, similar to that of B-cell lymphomas, than T-cell type tumors (P = 0.009). Among the 21 tumors, the prognosis of the four subtypes did not differ significantly; however, cases receiving aggressive chemotherapy and stem cell transplantation had a more favorable prognosis than those receiving only traditional chemotherapy and radiation therapy (P = 0.0089).
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
CD3 (MRQ-39) Rabbit Monoclonal Antibody