Merck
The influence of angiotensin-(1-7) Mas receptor agonist (AVE 0991) on mitochondrial proteome in kidneys of apoE knockout mice.
Biochimica et biophysica acta (2013-08-31)
Maciej Suski, Rafał Olszanecki, Aneta Stachowicz, Józef Madej, Beata Bujak-Giżycka, Krzysztof Okoń, Ryszard Korbut
RÉSUMÉ
Excessive action of angiotensin II on mitochondria has been shown to play an important role in mitochondrial dysfunction, a common feature of atherogenesis and kidney injury. Angiotensin-(1-7)/Mas receptor axis constitutes a countermeasure to the detrimental effects of angiotensin II on AT1 receptors. The aim of the study was to assess the effects of angiotensin-(1-7) peptidomimetic AVE0991 on the kidney mitochondrial proteome in widely used animal model of atherosclerosis (apoE(-/-) mice). Proteins changed in apoE(-/-) mice belonged to the groups of antioxidant enzymes, apoptosis regulators, inflammatory factors and metabolic enzymes. Importantly, AVE0991 partially reversed atherosclerosis-related changes in apoE(-/-) mice.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Trypsine from porcine pancreas, Proteomics Grade, BioReagent, Dimethylated