Merck
Efficient and improved synthesis of Telmisartan.
Beilstein journal of organic chemistry (2010-05-27)
A Sanjeev Kumar, Samir Ghosh, G N Mehta
RÉSUMÉ
An efficient synthesis of the angiotensin II receptor antagonist Telmisartan (1) is presented involving a cross coupling of 4-formylphenylboronic acid 10 with 2-(2-bromophenyl)-4,4-dimethyl-2-oxazoline (11) as the key step (90% yield). The benzimidazole moiety 15 was constructed regioselectively via a reductive amination-condensation sequence, replacing the alkylation of the preformed benzimidazole step in the previously published route. This methodology overcomes many of drawbacks associated with previously reported syntheses.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
N-Methyl-1,2-phenylenediamine, 97%