Merck
Niemann-pick type C2 deficiency in human fibroblasts confers robust and selective activation of prostaglandin E2 biosynthesis.
The Journal of biological chemistry (2013-07-03)
Andrey Frolov, Hua Dong, Min Jiang, Lihua Yang, Erik C Cook, Rahul Matnani, Bruce D Hammock, Leslie J Crofford
RÉSUMÉ
Activated fibroblasts, also known as myofibroblasts, are mediators of several major human pathologies including proliferative fibrotic disorders, invasive tumor growth, rheumatoid arthritis, and atherosclerosis. We previously identified Niemann-Pick type C2 (NPC2) protein as a negative regulator of fibroblast activation (Csepeggi, C., Jiang, M., Kojima, F., Crofford, L. J., and Frolov, A. (2011) J. Biol. Chem. 286, 2078-2087). Here we report that NPC2-deficiency leads to a dramatic up-regulation of the arachidonic acid (AA) metabolic pathway in human fibroblasts. The major enzymes in this pathway, cPLA2 type IVA, COX-2, and mPGES-1, were dramatically up-regulated at both the transcriptional and translational levels. The specific phenotypic changes resulted in a >10-fold increase in the production and secretion of a key modulator of inflammation and immunity, prostaglandin E2. More importantly, AA metabolome profiling by liquid chromatography/tandem mass-spectrometry revealed the very specific nature of prostaglandin E2 up-regulation as the other analyzed AA metabolites derived from the COX-2, cytochrome P450, 5/15-lipoxygenase, and non-enzymatic oxidative pathways were mostly down-regulated. Blocking activity of cPLA2 efficiently suppressed expression of inflammatory cytokines, IL-1β and IL-6, thereby identifying cPLA2 as an important regulator of the inflammatory program in NPC2-null cells. Altogether, these studies highlight NPC2 as a specific regulator of AA metabolism and inflammation that suggests potential for NPC2 protein or its related signaling in the treatment of inflammatory diseases characterized by the presence of activated fibroblasts.