Merck
ARNTL2 and SERPINE1: potential biomarkers for tumor aggressiveness in colorectal cancer.
Journal of cancer research and clinical oncology (2011-12-27)
Gianluigi Mazzoccoli, Valerio Pazienza, Anna Panza, Maria Rosa Valvano, Giorgia Benegiamo, Manlio Vinciguerra, Angelo Andriulli, Ada Piepoli
RÉSUMÉ
Cathepsin and plasmin may favor cancer cell invasion degrading extracellular matrix. Plasmin formation from plasminogen is regulated by plasminogen activator inhibitor type-1 (PAI-1). ARNTL2 activates the promoters of the PAI-1 gene, officially called SERPINE1, driving the circadian variation in circulating PAI-1 levels. We evaluated ARNTL2 and SERPINE1 expression in 50 colorectal cancer specimens and adjacent normal tissue and in colon cancer cell lines. We found up-regulation of ARNTL2 (P = 0.004) and SERPINE1 (P = 0.002) in tumor tissue. A statistically significant association was found between high ARNTL2 mRNA levels and vascular invasion (P < 0.0001), and between high SERPINE1 mRNA levels and microsatellite instability (MSI-H and MSI-L, P = 0.025). Sorting the subjects into quartile groups, a statistically significant association was found between high ARNTL2 expression and lymph node involvement (P < 0.001), between high SERPINE1 expression and grading (P < 0.001) and between high SERPINE1 expression and MSI H-L (P < 0.0001). In SW480 cells, a more proliferative model compared to CaCo2 cells, there were higher mRNA levels of ARNTL2 (P < 0.001) and SERPINE1 (P = 0.001). ARNTL2 and SERPINE1 expression is increased in colorectal cancer and in a highly proliferative colon cancer cell line and is related to tumor invasiveness and aggressiveness.