Merck
Heat shock protein-peptide complexes in cancer immunotherapy.
Current opinion in immunology (1994-10-01)
P K Srivastava, H Udono
RÉSUMÉ
Heat shock proteins (HSPs) are associated with a broad spectrum of peptides derived from the cells from which they are isolated. Vaccination with such HSP-peptide complexes elicits protective immunity against tumors or other cells used as the source of HSPs. These observations suggest that HSP-peptide complexes are suitable as vaccines against cancers and infectious diseases.