Merck
Preparation and characterization of nanoporous thin films from fully aliphatic polyimides.
Journal of nanoscience and nanotechnology (2011-11-30)
Hwan-Chul Yu, S Vijay Kumar, Sung-Youl Cho, Kazuaki Kudo, Toshio Takayama, Jae-Min Oh, Huihun Jung, Joong-Gon Kim, Chan-Moon Chung
RÉSUMÉ
Fully aliphatic polyimides (APIs) were prepared from rel-(1'R,3S5'S)-spiro[furan-3(2H),6'-[3]oxabicyclo[3.2.1]octane]-2,2',4',5(4H)-tetrone (DAn) as unsymmetrical spiro dianhydride, and either cis-trans-1,4-diaminocyclohexane (mix-DACH) or trans-1,4-diaminocyclohexane (trans-DACH) as diamine. Structure of all prepared monomers and polymers was confirmed via 1H-NMR and FT-IR. The solubility, optical transparency, and thermal properties of the full APIs were investigated. The solubility and decomposition temperature of the full APIs were found to be correlated with their intermolecular regularity confirmed via wide-angle X-ray diffraction (WAXD). Triblock copolyimides were synthesized through the incorporation of a thermally labile polymer, poly(propylene glycol) (PPG), into the full APIs, and their thermal properties were studied via thermogravimetric analysis (TGA). Nanoporous thin films of the full APIs were prepared via thermolysis of the labile block in the copolyimide films. Phase separation and nanopore formation in the copolymer films were confirmed via atomic force microscopy (AFM) and scanning electron microscopy (SEM), respectively. Nanoporous pores were successfully prepared inside the films.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
trans-1,4-Diaminocyclohexane, ≥98.0% (GC)