Merck
Erythema-multiforme-like, urticarial papular and plaque eruptions from bufexamac: report of 4 cases.
Contact dermatitis (1994-08-01)
P Koch, F A Bahmer
RÉSUMÉ
4 patients developed an erythema-multiforme-like reaction following acute contact dermatitis from 2 different bufexamac-containing topical preparations. Histologically, the lesions did not show changes that were typical of erythema multiforme. Patch testing gave positive reactions to bufexamac at 1, 2, 3 and 4 days. The histological picture of 2 positive test reactions was suggestive of a drug reaction in the 1st, and of an acute eczematous reaction in the 2nd, case. A systemic allergic reaction with spread of lesions could have been involved, following percutaneous absorption of the allergen.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Bufexamac, European Pharmacopoeia (EP) Reference Standard