Merck
Alkyne-functionalized ruthenium nanoparticles: ruthenium-vinylidene bonds at the metal-ligand interface.
Journal of the American Chemical Society (2012-01-11)
Xiongwu Kang, Nathaniel B Zuckerman, Joseph P Konopelski, Shaowei Chen
RÉSUMÉ
Stable ruthenium nanoparticles were prepared by the self-assembly of 1-dodecyne onto the "bare" Ru colloid surface. The formation of a Ru-vinylidene (Ru═C═CH-R) interfacial bonding linkage was confirmed by the specific reactivity of the nanoparticles with imine derivatives to form a heterocyclic complex at the metal-ligand interface, as manifested in (1)H and (13)C NMR, photoluminescence, and electrochemical measurements in which a ferrocenyl imine was used as the labeling probe. Notably, the resulting nanoparticles could also undergo olefin metathesis reactions with vinyl-terminated molecules, as exemplified by the functionalization of the nanoparticles with 1-vinylpyrene. In sharp contrast, no reactvity was observed with 1-dodecynide-stabilized ruthenium nanoparticles with either imine or vinyl derivatives, indicating that these (deprotonated) nanoparticles were stabilized instead by the formation of a Ru-C≡ dπ bond at the metal-ligand interface.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Dodecyne, 98%