Merck
Nucleation of an oil phase in a nonionic microemulsion-containing chlorinated oil upon systematic temperature quench.
The journal of physical chemistry. B (2010-05-25)
G Roshan Deen, Jan Skov Pedersen
RÉSUMÉ
A clear and stable nonionic model microemulsion consisting of pentaoxyethylene glycol dodecyl ether (C(12)E(5)), water, and 1-chlorotetradecane (CLTD) was prepared. This system was subjected to a systematic temperature quench (perturbation out of equilibrium) in steps of 1.0 degrees C from 20.4 to 15.3 degrees C in the unstable region of its phase diagram. The change in turbidity (for droplet volume fractions of 0.02 and 0.08) and hydrodynamic radius (R(h)) (for a droplet volume fraction of 0.02) of the system on its way to its new equilibrium was measured at each quench temperature. For small systematic temperature quenches just below the emulsification failure boundary (EFB) the turbidity decreases and remains constant indicating quick changes in the microstructures. Further lowering of temperature brings the system to the unstable region where the turbidity and light scattering increase sharply as function of time because of expulsion of excess oil from the microemulsion droplets. The newly formed oil-rich droplets grow in size as a function of time. These observations indicate the existence of a narrow but observable metastable region en route to the new equilibrium where both microemulsion droplets and larger oil-rich droplets coexist. The region in which microemulsion droplets are metastable is very narrow and is concentration-dependent. The presence of a metastable region is as for other similar systems attributed to the presence of a free energy barrier for the formation of the larger oil-rich droplets associated with curvature free energy of the surfactant film. The turbidity-time curves were converted to the radius-time curves using a model assuming monodisperse spherical droplets. The obtained results are in good agreement with the results for the hydrodynamic radius. The observed average radius from both type of measurements decreases in the metastable region. By performing calculation of the influence of eccentricity and size polydispersity on the observed radius, we have shown that the distribution of the microemulsion droplets becomes more homogeneous in the metastable region.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Chlorotetradecane, 98%