Merck
  • Lewis acid-promoted reactions of zirconacyclopentadienes with isocyanates. A one-pot three-component synthesis of multiply-substituted iminocyclopentadienes from one isocyanate and two alkynes.
Lewis acid-promoted reactions of zirconacyclopentadienes with isocyanates. A one-pot three-component synthesis of multiply-substituted iminocyclopentadienes from one isocyanate and two alkynes.
Chemical communications (Cambridge, England) (2005-09-30)
Jiang Lu, Guoliang Mao, Wenxiong Zhang, Zhenfeng Xi
RÉSUMÉ
Multiply-substituted iminocyclopentadienes were formed from Lewis acid-promoted reactions of zirconacyclopentadienes and isocyanates via a one-pot three-component coupling process; the C=O double bond of the RN=C=O moiety in the isocyanate was cleaved, and the isocyanates behaved formally as a one-carbon unit with Lewis acid-dependent and substituent-dependent reactions being realized.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Bromophenyl isocyanate, 99%